Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021537.3 | NP_067512.3 | 8 | 1060 | ||
GenBank mRNA | AF004934.1 | - | 7 | 898 | ||
AK148041.1 | - | 8 | 1034 | |||
AK152917.1 | - | 8 | 1003 | |||
BC029875.1 | - | 7 | 1778 | |||
BC052913.1 | - | 8 | 995 | 30057101 | ||
BC071218.1 | - | 8 | 1055 | 4012695 |