Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025359.3 | NP_079635.1 | 2 | 333 | ||
GenBank mRNA | AK008175.1 | - | 2 | 273 | ||
AK012571.1 | - | 2 | 345 | |||
AK090217.1 | - | 2 | 345 | |||
AK090322.1 | - | 2 | 345 | |||
AK158761.1 | - | 2 | 338 | |||
AK165768.1 | - | 2 | 370 | |||
AK167495.1 | - | 2 | 385 | |||
AK167979.1 | - | 2 | 338 | |||
BC018317.1 | - | 2 | 333 | 4166625 |