Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025383.3 | NP_079659.1 | 5 | 459 | ||
GenBank mRNA | AK003463.1 | - | 5 | 426 | ||
AK145874.1 | - | 5 | 447 | |||
AK155871.1 | - | 5 | 447 | |||
AK156566.1 | - | 5 | 436 | |||
AK170987.1 | - | 5 | 444 | |||
AK172538.1 | - | 5 | 430 | |||
AK185656.1 | - | 1 | 63 | |||
AK188919.1 | - | 2 | 62 | |||
AK207758.1 | - | 2 | 62 | |||
AK212453.1 | - | 2 | 62 | |||
AK213306.1 | - | 2 | 62 | |||
AK213654.1 | - | 2 | 62 | |||
BC037069.1 | - | 5 | 458 | 5002765 |