Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175112.5 | NP_780321.1 | 12 | 1350 | ||
GenBank mRNA | AK014325.1 | - | 12 | 1306 | ||
AK017746.1 | - | 12 | 1271 | |||
AK079383.1 | - | 12 | 1331 | |||
AK146058.1 | - | 12 | 1315 | |||
AK156681.1 | - | 12 | 1315 | |||
AK177228.1 | - | 2 | 267 | |||
AK217457.1 | - | 3 | 337 | |||
BC059051.1 | - | 12 | 1272 | 6818771 | ||
BC060072.1 | - | 10 | 3811 | 5718934 | ||
CO041965.1 | - | 1 | 402 |