Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrasls5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025731.2 | NP_080007.1 | 6 | 884 | ||
GenBank mRNA | AB261168.1 | - | 6 | 842 | ||
AB447517.1 | - | 6 | 842 | |||
AK006527.1 | - | 6 | 849 | |||
AK014971.1 | - | 6 | 885 | |||
BC061008.1 | - | 6 | 864 | 6772458 |