Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039965.1 | NP_001035054.1 | 5 | 1944 | ||
NM_001039967.1 | NP_001035056.1 | 6 | 2052 | |||
NM_181274.3 | NP_851791.1 | 5 | 2012 | |||
GenBank mRNA | AK004570.1 | - | 5 | 1955 | ||
AK030679.1 | - | 6 | 2050 | |||
AK054059.1 | - | 1 | 1298 | |||
AK077321.1 | - | 1 | 125 | |||
AK083392.1 | - | 6 | 1967 | |||
AK088889.1 | - | 4 | 1855 | |||
AK136878.1 | - | 5 | 1948 | |||
BC063330.1 | - | 2009 | ||||
BC079868.1 | - | 7 | 2547 |