Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 7 | 1091 | ||
GenBank mRNA | AB086655.1 | - | 7 | 1070 | ||
AF477481.1 | - | 7 | 1091 | |||
AK012041.1 | - | 7 | 1068 | |||
AK017609.1 | - | 7 | 1065 | |||
AK028287.1 | - | 7 | 1063 | |||
AK049163.1 | - | 7 | 1063 | |||
AK076527.1 | - | 6 | 1173 | |||
AK083015.1 | - | 7 | 1066 | |||
BC013842.1 | - | 7 | 1046 | |||
BC024485.1 | - | 4 | 342 | |||
BC048076.1 | - | 7 | 1091 | 6307476 |