Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026290.3 | NP_080566.2 | 3 | 344 | ||
GenBank mRNA | AK005672.1 | - | 3 | 341 | ||
AK015756.1 | - | 3 | 344 | |||
BC049559.1 | - | 3 | 343 | 6707229 | ||
EU477370.1 | - | 3 | 321 |