Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026395.1 | NP_080671.1 | 2 | 149 | ||
GenBank mRNA | AK002550.1 | - | 2 | 149 | ||
AK003682.1 | - | 2 | 171 | |||
AK013262.1 | - | 2 | 206 | |||
AK078396.1 | - | 2 | 212 | |||
AK168192.1 | - | 3 | 368 | |||
AK178378.1 | - | 2 | 277 | |||
AK178715.1 | - | 2 | 277 | |||
AK182182.1 | - | 2 | 279 | |||
BC029189.1 | - | 1 | 55 | 4036721 |