Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023409.4 | NP_075898.1 | 4 | 676 | ||
GenBank mRNA | AB021289.1 | - | 4 | 431 | ||
AK008603.1 | - | 4 | 463 | |||
AK009127.1 | - | 4 | 493 | |||
AK012239.1 | - | 4 | 523 | |||
AK149805.1 | - | 4 | 466 | |||
AK151067.1 | - | 4 | 463 | |||
AK151080.1 | - | 2 | 1888 | |||
AK151133.1 | - | 4 | 464 | |||
AK152236.1 | - | 4 | 463 | |||
AK153020.1 | - | 4 | 463 | |||
AK153203.1 | - | 4 | 463 | |||
AK154838.1 | - | 4 | 463 | |||
AK158624.1 | - | 4 | 644 | |||
AK159154.1 | - | 4 | 510 | |||
AK165263.1 | - | 4 | 489 | |||
AK167869.1 | - | 4 | 464 | |||
AK170327.1 | - | 4 | 464 | |||
BC003471.1 | - | 4 | 478 | 2649564 | ||
BC007190.1 | - | 4 | 510 | 3592515 |