Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023409.4 | NP_075898.1 | 3 | 582 | ||
GenBank mRNA | AB021289.1 | - | 3 | 337 | ||
AK008603.1 | - | 3 | 369 | |||
AK009127.1 | - | 3 | 399 | |||
AK012239.1 | - | 3 | 429 | |||
AK054441.1 | - | 1 | 1377 | |||
AK149805.1 | - | 3 | 372 | |||
AK151067.1 | - | 3 | 369 | |||
AK151080.1 | - | 1 | 1794 | |||
AK151133.1 | - | 3 | 370 | |||
AK152236.1 | - | 3 | 369 | |||
AK153020.1 | - | 3 | 369 | |||
AK153203.1 | - | 3 | 369 | |||
AK154838.1 | - | 3 | 369 | |||
AK158624.1 | - | 3 | 550 | |||
AK159154.1 | - | 3 | 416 | |||
AK165263.1 | - | 3 | 395 | |||
AK167869.1 | - | 3 | 370 | |||
AK170327.1 | - | 3 | 370 | |||
BC003471.1 | - | 3 | 384 | 2649564 | ||
BC007190.1 | - | 3 | 416 | 3592515 |