Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fitm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026808.1 | NP_081084.1 | 2 | 731 | ||
GenBank mRNA | AB054000.1 | - | 2 | 731 | ||
AK003939.1 | - | 2 | 620 | |||
BC037188.1 | - | 2 | 752 | |||
BC048823.1 | - | 2 | 529 | |||
BC104408.1 | - | 2 | 731 | 40050546 | ||
BC104409.2 | - | 2 | 731 | 40050547 |