Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164819.1 | NP_001158291.1 | 2 | 360 | ||
NM_001164820.1 | NP_001158292.1 | 2 | 360 | |||
NM_027172.3 | NP_081448.2 | 2 | 428 | |||
GenBank mRNA | AK009850.1 | - | 2 | 311 | ||
AK029091.1 | - | 2 | 405 | |||
AK047618.1 | - | 2 | 361 | |||
AK154970.1 | - | 2 | 410 | |||
AK170158.1 | - | 2 | 389 | |||
BC016127.1 | - | 2 | 329 | 4910227 |
Set Membership: |
![]() |