Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164819.1 | NP_001158291.1 | 3, 4 | 1186 | ||
NM_027172.3 | NP_081448.2 | 3, 4 | 1254 | |||
GenBank mRNA | AK009850.1 | - | 3, 4 | 1137 | ||
AK029091.1 | - | 3, 4 | 1231 | |||
AK047618.1 | - | 3, 4 | 1187 | |||
AK154970.1 | - | 3, 4 | 1236 | |||
AK170158.1 | - | 3, 4 | 1215 |
Set Membership: |
![]() |