Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253694.1 | NP_001240623.1 | 4 | 1266 | ||
NM_001253695.1 | NP_001240624.1 | 3 | 995 | |||
NM_001253696.1 | NP_001240625.1 | 2 | 1064 | |||
NM_027248.2 | NP_081524.2 | 3 | 1153 | |||
GenBank mRNA | AB063578.1 | - | 3 | 1153 | ||
AK008516.1 | - | 1 | 471 | |||
AK053445.1 | - | 3 | 991 | |||
AK154332.1 | - | 4 | 1241 | |||
BC034874.1 | - | 3 | 1030 | |||
BC052050.1 | - | 4 | 1039 | |||
BC071271.1 | - | 2 | 924 | 30290129 | ||
BC096486.1 | - | 2 | 1032 | 5352045 |