Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153551.1 | NP_705779.1 | 13 | 957 | ||
GenBank mRNA | AK040537.1 | - | 13 | 975 | ||
BC035208.1 | - | 13 | 957 | 3671384 |