Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027633.3 | NP_081909.2 | 3 | 492 | ||
GenBank mRNA | AK016461.1 | - | 3 | 433 | ||
BC042706.1 | - | 1 | 148 | |||
BC049660.1 | - | 3 | 492 | 6742399 |