Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081490.2 | NP_001074959.1 | 5 | 411 | ||
NM_023605.2 | NP_076094.2 | 5 | 713 | |||
GenBank mRNA | AB041585.1 | - | 5 | 682 | ||
AK018482.1 | - | 5 | 664 | |||
AK077607.1 | - | 5 | 374 | |||
AK182638.1 | - | 4 | 303 | |||
BC020074.1 | - | 5 | 684 | 3710611 | ||
DV657928.1 | - | 5 | 399 |