Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028627.2 | NP_082903.2 | 9 | 2118 | ||
GenBank mRNA | AK220438.1 | - | 4 | 2235 | ||
BC043135.1 | - | 5 | 947 | |||
BC057353.1 | - | 9 | 2119 | |||
BC138652.1 | - | 7 | 1977 | 8861672 | ||
BC145352.1 | - | 7 | 1980 | 9053908 |