Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028627.2 | NP_082903.2 | 14, 15 | 2605 | ||
GenBank mRNA | AK158685.1 | - | 4, 5 | 411 | ||
AK158851.1 | - | 4, 5 | 537 | |||
AK220438.1 | - | 9, 10 | 2722 | |||
BC043135.1 | - | 10, 11 | 1434 | |||
BC057353.1 | - | 14, 15 | 2606 | |||
BC138652.1 | - | 12, 13 | 2464 | 8861672 | ||
BC145352.1 | - | 12, 13 | 2467 | 9053908 |