Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029285.1 | NP_083561.1 | 1 | 60 | ||
GenBank mRNA | AK005574.1 | - | 1 | 60 | ||
AK006710.1 | - | 1 | 93 | |||
BC060996.1 | - | 1 | 63 | 6743526 |