Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029285.1 | NP_083561.1 | 4 | 552 | ||
GenBank mRNA | AK005574.1 | - | 4 | 552 | ||
BC060996.1 | - | 4 | 555 | 6743526 |