Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290569.1 | NP_001277498.1 | 7 | 815 | ||
NM_029377.2 | NP_083653.1 | 7 | 689 | |||
GenBank mRNA | AK007858.1 | - | 7 | 690 | ||
AY428767.1 | - | 7 | 539 | |||
BC013546.1 | - | 7 | 671 | 4210417 |