Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023824.3 | NP_076313.1 | 2 | 598 | ||
GenBank mRNA | AB041564.1 | - | 2 | 519 | ||
AK005142.1 | - | 2 | 621 | |||
AK033625.1 | - | 2 | 616 | |||
AK044092.1 | - | 2 | 418 | |||
AK044486.1 | - | 2 | 621 | |||
AK152676.1 | - | 2 | 597 | |||
AY424293.1 | - | 2 | 549 | |||
BC011185.1 | - | 2 | 598 | 4196033 | ||
BC026921.1 | - | 2 | 549 | 4500871 |