Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176838.2 | NP_789808.1 | 13 | 1978 | ||
GenBank mRNA | AK088755.1 | - | 13 | 1848 | ||
AK168488.1 | - | 12 | 1211 | |||
BC031444.1 | - | 13 | 1978 | 4222237 |