Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030150.2 | NP_084426.2 | 2 | 254 | ||
GenBank mRNA | AF316999.1 | - | 1 | 153 | ||
AK021012.1 | - | 2 | 254 | |||
AK153342.1 | - | 2 | 183 | |||
BC029209.1 | - | 1 | 164 | 2651254 | ||
BY727457.1 | - | 2 | 254 |