Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 2 | 387 | ||
AK002290.1 | - | 2 | 281 | |||
AK146657.1 | - | 2 | 349 | |||
AK158530.1 | - | 2 | 246 | |||
AK160924.1 | - | 2 | 283 | |||
AK164392.1 | - | 2 | 312 | |||
AK167718.1 | - | 2 | 286 | |||
AK168220.1 | - | 2 | 300 | |||
AK168291.1 | - | 2 | 300 | |||
AK187532.1 | - | 2 | 90 | |||
BC012962.1 | - | 2 | 285 | 4188579 | ||
BY251864.1 | - | 2 | 405 |