Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hecw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081348.3 | NP_001074817.3 | 7 | 1122 | ||
GenBank mRNA | AB083710.1 | - | 5 | 625 | ||
AK047678.1 | - | 7 | 1057 | |||
AK051569.1 | - | 6 | 943 | |||
AK053694.1 | - | 6 | 1067 | |||
AK084246.1 | - | 6 | 943 |