Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134020.1 | NP_598781.1 | 4 | 450 | ||
GenBank mRNA | AF368921.1 | - | 4 | 481 | ||
AK078425.1 | - | 4 | 461 | |||
AK158876.1 | - | 4 | 483 | |||
AK204643.1 | - | 2 | 145 | |||
BC023041.1 | - | 4 | 450 | 5360236 |