Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252482.1 | NP_001239411.1 | 5 | 604 | ||
NM_001252483.1 | NP_001239412.1 | 5 | 565 | |||
NM_001252484.1 | NP_001239413.1 | 5 | 495 | |||
NM_001252485.1 | NP_001239414.1 | 6 | 724 | |||
NM_213729.1 | NP_998894.1 | 5 | 633 | |||
GenBank mRNA | AK139002.1 | - | 5 | 493 | ||
AY601909.1 | - | 5 | 633 | |||
AY601910.1 | - | 5 | 604 | |||
AY601911.1 | - | 5 | 565 | |||
AY601912.1 | - | 5 | 495 | |||
BC096427.1 | - | 6 | 678 | 4160694 | ||
BC115572.1 | - | 5 | 464 | 40102153 | ||
BC115573.1 | - | 5 | 464 | 40102154 |