Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026924.3 | NP_081200.2 | 4 | 935 | ||
NM_152947.2 | NP_694455.2 | 4 | 676 | |||
GenBank mRNA | AB101294.1 | - | 4 | 838 | ||
AB101295.1 | - | 3 | 880 | |||
AB101296.1 | - | 4 | 1010 | |||
AK007360.1 | - | 3 | 561 | |||
AK167697.1 | - | 4 | 926 | |||
AY090537.1 | - | 4 | 746 | |||
AY090538.1 | - | 4 | 676 | |||
BC031771.1 | - | 4 | 935 | 4219296 | ||
BC057210.1 | - | 4 | 956 | 5329625 | ||
BC094445.1 | - | 4 | 947 | 4981833 |