Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026241.3 | NP_080517.1 | 4 | 451 | ||
GenBank mRNA | AK018681.1 | - | 4 | 452 | ||
AK030874.1 | - | 647 | ||||
AK042896.1 | - | 4 | 452 | |||
AK081207.1 | - | 4 | 644 | |||
AK158215.1 | - | 4 | 646 | |||
AK160231.1 | - | 4 | 467 | |||
BC117004.1 | - | 4 | 481 | 40126323 | ||
BC119100.1 | - | 4 | 481 | 8733849 | ||
BC144893.1 | - | 4 | 516 |