Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053267.2 | NP_444497.1 | 3 | 421 | ||
GenBank mRNA | AK018791.1 | - | 3 | 257 | ||
AK035312.1 | - | 3 | 211 | |||
AK046034.1 | - | 3 | 213 | |||
AK079358.1 | - | 3 | 239 | |||
AK079576.1 | - | 2 | 629 | |||
AW049879.1 | - | 3 | 464 | |||
AY043488.1 | - | 3 | 237 | |||
BC019742.1 | - | 3 | 113 | |||
BC147332.1 | - | 3 | 331 | 9056406 | ||
BC147333.1 | - | 3 | 333 | 9056396 | ||
BI731890.1 | - | 3 | 419 | |||
CK787580.1 | - | 3 | 413 |