Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007570.2 | NP_031596.1 | 1, 2 | 243 | ||
GenBank mRNA | AK088976.1 | - | 1, 2 | 206 | ||
AK133796.1 | - | 1, 2 | 208 | |||
AK169269.1 | - | 1, 2 | 208 | |||
AK169436.1 | - | 1, 2 | 208 | |||
BC132259.1 | - | 1, 2 | 193 | 40130536 | ||
BC138639.1 | - | 1, 2 | 193 | 8861659 | ||
BY766758.1 | - | 1, 2 | 248 |