Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163432.1 | NP_001156904.1 | 13 | 1653 | ||
NM_007712.3 | NP_031738.2 | 13 | 1656 | |||
GenBank mRNA | AF033564.1 | - | 13 | 1378 | ||
AK148377.1 | - | 11 | 2684 | |||
AK161259.1 | - | 9 | 1828 | |||
BC015080.1 | - | 13 | 1641 | 3995512 | ||
CD740936.1 | - | 1 | 502 |