Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 20, 21 | 2946 | ||
NM_001284248.1 | NP_001271177.1 | 22, 23 | 3458 | |||
NM_001284249.1 | NP_001271178.1 | 21, 22 | 3055 | |||
NM_001284250.1 | NP_001271179.1 | 20, 21 | 2943 | |||
NM_010412.3 | NP_034542.3 | 20, 21 | 2943 | |||
GenBank mRNA | AF006602.2 | - | 19, 20 | 2656 | ||
AF207748.1 | - | 19, 20 | 2575 | |||
AK122325.2 | - | 15, 16 | 1985 | |||
AK139202.1 | - | 21, 22 | 3029 | |||
AK145972.1 | - | 20, 21 | 2909 | |||
AK146483.1 | - | 20, 21 | 2882 | |||
AK155335.1 | - | 20, 21 | 2917 | |||
AK156008.1 | - | 14, 15 | 3780 | |||
AK157159.1 | - | 14, 15 | 3780 | |||
AK200348.1 | - | 2, 3 | 118 | |||
BC023309.1 | - | 11, 12 | 1496 | |||
BC049926.1 | - | 20, 21 | 2877 | |||
BC058554.1 | - | 22, 23 | 3410 | |||
BC060609.1 | - | 19, 20 | 2737 | 5720438 |