Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011113.3 | NP_035243.1 | 6, 7 | 767 | ||
GenBank mRNA | AK002580.1 | - | 6, 7 | 838 | ||
AK131951.1 | - | 6, 7 | 783 | |||
AK144796.1 | - | 6, 7 | 784 | |||
AK146827.1 | - | 6, 7 | 782 | |||
AK154141.1 | - | 6, 7 | 784 | |||
AK155351.1 | - | 6, 7 | 771 | |||
BC010309.1 | - | 5, 6 | 1285 | |||
X62700.1 | - | 6, 7 | 768 | |||
X62701.1 | - | 6, 7 | 986 |