Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042767.2 | NP_001036232.1 | 9 | 942 | ||
NM_001042768.2 | NP_001036233.2 | 10 | 1022 | |||
NM_008934.3 | NP_032960.3 | 9 | 954 | |||
GenBank mRNA | AF318182.1 | - | 9 | 931 | ||
AI118438.1 | - | 1 | 636 | |||
BC013896.1 | - | 9 | 959 | 4211329 | ||
CF550816.1 | - | 5 | 518 | |||
D10445.1 | - | 8 | 870 |