Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011226.1 | NP_035356.1 | 3, 4 | 490 | ||
GenBank mRNA | AB232613.1 | - | 2, 3 | 396 | ||
AK017590.1 | - | 3, 4 | 524 | |||
BC032936.1 | - | 3, 4 | 490 | 1379043 | ||
X80473.1 | - | 2, 3 | 396 |