Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009144.2 | NP_033170.1 | 3 | 926 | ||
GenBank mRNA | AF017989.1 | - | 3 | 958 | ||
AK138252.1 | - | 3 | 765 | |||
AK147091.1 | - | 3 | 923 | |||
BC014722.1 | - | 3 | 886 | 4487469 | ||
D50462.1 | - | 3 | 718 | |||
U88567.1 | - | 3 | 940 |
Set Membership: |
G-protein coupled receptor |