Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009144.2 | NP_033170.1 | 1 | 325 | ||
GenBank mRNA | AK138252.1 | - | 1 | 164 | ||
AK147091.1 | - | 1 | 322 | |||
BC014722.1 | - | 1 | 285 | 4487469 | ||
D50462.1 | - | 1 | 117 | |||
U88567.1 | - | 1 | 339 |
Set Membership: |
![]() |