Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013678.2 | NP_038706.1 | 6 | 755 | ||
GenBank mRNA | AK004931.1 | - | 2 | 2828 | ||
AK157125.1 | - | 6 | 769 | |||
AK165559.1 | - | 6 | 762 | |||
AK172018.1 | - | 6 | 755 | |||
AK215042.1 | - | 1 | 21 | |||
BC049779.1 | - | 5 | 2086 | |||
BC096401.1 | - | 6 | 755 | 1365532 |