Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 5 | 311 | ||
GenBank mRNA | AK012359.1 | - | 5 | 309 | ||
AK078555.1 | - | 5 | 315 | |||
AK150504.1 | - | 5 | 337 | |||
AK150627.1 | - | 5 | 337 | |||
AK150940.1 | - | 5 | 329 | |||
AK152274.1 | - | 5 | 309 | |||
AK159561.1 | - | 5 | 546 | |||
AK168199.1 | - | 5 | 282 | |||
AK168328.1 | - | 5 | 308 | |||
BC003731.1 | - | 5 | 311 | 3601668 | ||
U30838.1 | - | 5 | 311 |