Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271443.1 | NP_001258372.1 | 4 | 251 | ||
NM_001271444.1 | NP_001258373.1 | 4 | 491 | |||
NM_144870.5 | NP_659119.2 | 4 | 494 | |||
GenBank mRNA | AK135069.1 | - | 4 | 252 | ||
AK149674.1 | - | 4 | 452 | |||
AK153182.1 | - | 4 | 438 | |||
AK153900.1 | - | 4 | 228 | |||
AK164901.1 | - | 4 | 228 | |||
AK167907.1 | - | 4 | 229 | |||
BC021616.1 | - | 4 | 212 | 5132866 | ||
BC086766.1 | - | 4 | 205 | 6820786 |