Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yrdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153566.2 | NP_705794.2 | 1, 2 | 408 | ||
GenBank mRNA | AK153627.1 | - | 1, 2 | 408 | ||
AY283537.1 | - | 1, 2 | 384 | |||
BC023823.1 | - | 1, 2 | 394 | 5343817 |