Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146901.2 | NP_667112.1 | 1 | 340 | ||
GenBank mRNA | BC132097.1 | - | 1 | 340 | 40130374 | |
BC138391.1 | - | 1 | 340 | 8861411 |