Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009940.1 | NP_001009940.1 | 1, 2 | 149 | ||
GenBank mRNA | AF453945.1 | - | 1, 2 | 149 | ||
BC115793.1 | - | 2, 3 | 208 | 40100999 |