Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 17, 18 | 2158 | ||
NM_026082.5 | NP_080358.3 | 17, 18 | 2158 | |||
GenBank mRNA | BC094420.1 | - | 4, 5 | 354 | ||
CA752561.1 | - | 4, 5 | 356 |