Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU022855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080354.1 | - | 1 | 1503 | ||
AU022855.1 | - | 1 | 283 |
Gene Symbol: A630075K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042254.1 | - | 2217 |
Gene Symbol: Sgip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043905.1 | - | 1071 | |||
AK090117.1 | - | 1345 |