Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001184.1 | NP_001001184.1 | 13 | 1495 | ||
GenBank mRNA | AK030772.1 | - | 13 | 1509 | ||
AK036600.1 | - | 12 | 2178 | |||
BC065112.1 | - | 13 | 1495 | 30532532 |